Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019826.3 | NP_062800.1 | 4 | 461 | ||
GenBank mRNA | AF225989.1 | - | 4 | 440 | ||
AK005024.1 | - | 4 | 480 | |||
AK017660.1 | - | 4 | 447 | |||
AK077416.1 | - | 4 | 467 | |||
BC018325.1 | - | 4 | 451 | 4192286 | ||
BC027198.1 | - | 4 | 461 | 4237982 |