Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135694.2 | NP_001129166.1 | 9 | 812 | ||
NM_005662.6 | NP_005653.3 | 8 | 809 | |||
XM_006716394.1 | XP_006716457.1 | 7 | 896 | |||
GenBank mRNA | AF038962.1 | - | 8 | 698 | ||
AK058201.1 | - | 8 | 683 | |||
BC056870.1 | - | 8 | 694 | 6163890 | ||
U90943.1 | - | 6 | 599 |