Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019874.3 | NP_063927.1 | 1 | 227 | ||
GenBank mRNA | AF088983.1 | - | 2 | 274 | ||
AF321322.1 | - | 2 | 267 | |||
AK158972.1 | - | 2 | 527 | |||
BC027782.1 | - | 1 | 2970 | |||
BC048902.1 | - | 2 | 512 | 6396037 | ||
BC057087.1 | - | 1 | 227 | 5695243 |