Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019748.3 | NP_062722.1 | 9 | 1698 | ||
GenBank mRNA | AB024303.1 | - | 9 | 1586 | ||
AK010313.1 | - | 9 | 1575 | |||
AK011783.1 | - | 6 | 1045 | |||
AK087556.1 | - | 9 | 1589 | |||
AK090012.1 | - | 9 | 1585 | |||
AK159672.1 | - | 9 | 1600 | |||
BC068164.1 | - | 9 | 1587 | 30359189 |