Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019765.5 | NP_062739.2 | 9 | 2356 | ||
GenBank mRNA | AK014540.1 | - | 9 | 2127 | ||
BC007191.1 | - | 9 | 2296 | 3592614 |