Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019766.4 | NP_062740.1 | 8 | 983 | ||
GenBank mRNA | AB024935.1 | - | 8 | 791 | ||
AK008805.1 | - | 8 | 1014 | |||
AK010636.1 | - | 8 | 978 | |||
AK075932.1 | - | 8 | 850 | |||
AK075987.1 | - | 8 | 894 | |||
AK077538.1 | - | 8 | 984 | |||
AK088321.1 | - | 1 | 65 | |||
AK167443.1 | - | 8 | 957 | |||
AK168073.1 | - | 8 | 893 | |||
AK168210.1 | - | 8 | 893 | |||
AK168721.1 | - | 8 | 833 | |||
AY281130.1 | - | 8 | 1010 | |||
BC003708.1 | - | 8 | 874 | 3489418 | ||
BC085264.1 | - | 8 | 880 | 6392001 |