Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019776.2 | NP_062750.2 | 9, 10 | 1263 | ||
GenBank mRNA | AB021491.1 | - | 9, 10 | 1220 | ||
AK034914.1 | - | 8, 9 | 1132 | |||
AK088028.1 | - | 9, 10 | 1222 | |||
AK133837.1 | - | 9, 10 | 1269 | |||
AK159837.1 | - | 9, 10 | 1228 | |||
AK161591.1 | - | 9, 10 | 1217 | |||
AK162432.1 | - | 9, 10 | 1204 | |||
AK167576.1 | - | 9, 10 | 1253 | |||
AK190012.1 | - | 2, 3 | 131 | |||
AK196687.1 | - | 2, 3 | 131 | |||
AK200045.1 | - | 2, 3 | 131 | |||
AK216953.1 | - | 2, 3 | 131 | |||
BC007126.1 | - | 9, 10 | 1210 | 3592548 |