Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252543.1 | NP_001239472.1 | 5 | 669 | ||
NM_001252544.1 | NP_001239473.1 | 5 | 662 | |||
NM_001252545.1 | NP_001239474.1 | 5 | 651 | |||
NM_001281816.1 | NP_001268745.1 | 2 | 291 | |||
NM_019787.4 | NP_062761.2 | 5 | 680 | |||
GenBank mRNA | AF200326.1 | - | 4 | 552 | ||
AK014597.1 | - | 5 | 630 | |||
AK029661.1 | - | 2 | 292 | |||
AK139729.1 | - | 5 | 594 | |||
AK146083.1 | - | 5 | 629 | |||
AK149693.1 | - | 5 | 609 | |||
AK153214.1 | - | 5 | 636 | |||
AK155168.1 | - | 5 | 610 | |||
AK158398.1 | - | 5 | 647 | |||
AK166330.1 | - | 5 | 612 | |||
AK167382.1 | - | 5 | 618 | |||
AK167732.1 | - | 5 | 598 | |||
AK171597.1 | - | 5 | 610 | |||
BC005464.1 | - | 5 | 605 | 3591420 | ||
BC011160.1 | - | 5 | 632 | 4024353 |