Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vapb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019806.5 | NP_062780.2 | 6 | 885 | ||
GenBank mRNA | AK050632.1 | - | 6 | 834 | ||
AK081908.1 | - | 6 | 814 | |||
AK160925.1 | - | 6 | 728 | |||
AK172080.1 | - | 6 | 658 | |||
AK205322.1 | - | 1 | 27 | |||
BC024562.1 | - | 1 | 35 | |||
BC048231.1 | - | 6 | 885 | 6313869 | ||
BC057123.1 | - | 6 | 874 | 6411688 |