Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019822.3 | NP_062796.2 | 9, 10 | 1224 | ||
GenBank mRNA | AF225959.1 | - | 9, 10 | 1140 | ||
AK010919.1 | - | 9, 10 | 1195 | |||
AK075674.1 | - | 7, 8 | 847 | |||
AK145474.1 | - | 9, 10 | 1186 | |||
AK145792.1 | - | 9, 10 | 1215 | |||
AK150091.1 | - | 9, 10 | 1178 | |||
AK166878.1 | - | 9, 10 | 1213 | |||
AK178559.1 | - | 19 | ||||
AK190773.1 | - | 19 | ||||
AK210109.1 | - | 19 | ||||
AK219626.1 | - | 19 | ||||
BC008974.1 | - | 9, 10 | 1224 | 3586698 | ||
BC019746.1 | - | 7, 8 | 859 | 4188750 |