Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019879.3 | NP_063932.2 | 2 | 343 | ||
GenBank mRNA | AF144101.2 | - | 2 | 176 | ||
AK002754.1 | - | 2 | 178 | |||
AK005080.1 | - | 2 | 192 | |||
AK087390.1 | - | 2 | 181 | |||
AK139288.1 | - | 2 | 171 | |||
AK184184.1 | - | 2 | 119 | |||
BC011087.1 | - | 2 | 164 | 4012310 | ||
BY061062.1 | - | 2 | 344 |