Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301671.1 | NP_001288600.1 | 3 | 522 | ||
NM_001301672.1 | NP_001288601.1 | 3 | 396 | |||
NM_001301673.1 | NP_001288602.1 | 3 | 405 | |||
NM_019929.4 | NP_064313.1 | 3 | 396 | |||
GenBank mRNA | AA105565.1 | - | 2 | 125 | ||
AF063847.1 | - | 3 | 223 | |||
AK012742.1 | - | 3 | 271 | |||
AK013019.1 | - | 3 | 265 | |||
AK148306.1 | - | 3 | 260 | |||
AK150063.1 | - | 3 | 269 | |||
AK150200.1 | - | 3 | 269 | |||
AK160169.1 | - | 3 | 268 | |||
AK162035.1 | - | 3 | 269 | |||
AK167207.1 | - | 3 | 237 | |||
AK168988.1 | - | 3 | 260 | |||
AK170896.1 | - | 3 | 454 | |||
AK171169.1 | - | 3 | 454 | |||
AK186890.1 | - | 2 | 108 | |||
BC115488.1 | - | 3 | 203 | 40100722 | ||
BC115489.1 | - | 3 | 203 | 40100723 | ||
CJ069985.1 | - | 3 | 307 | |||
CJ159246.1 | - | 3 | 313 | |||
CK619326.1 | - | 3 | 256 |