Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042543.2 | NP_001036008.1 | 9 | 2091 | ||
NM_006529.3 | NP_006520.2 | 10 | 2136 | |||
XM_011532268.2 | XP_011530570.1 | 6 | 1382 | |||
XM_017008630.1 | XP_016864119.1 | 11 | 2160 | |||
GenBank mRNA | AF018157.1 | - | 4 | 899 | ||
BC036086.1 | - | 10 | 2135 | 5286731 | ||
U93917.1 | - | 9 | 2007 |