Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177323.2 | NP_001170794.1 | 2 | 409 | ||
NM_001177324.1 | NP_001170795.1 | 2 | 412 | |||
NM_001253706.1 | NP_001240635.1 | 2 | 412 | |||
NM_019942.5 | NP_064326.2 | 2 | 412 | |||
GenBank mRNA | AB023622.1 | - | 2 | 176 | ||
AK012858.1 | - | 3 | 206 | |||
AK048455.1 | - | 2 | 234 | |||
AK083302.1 | - | 2 | 212 | |||
AK088406.1 | - | 2 | 192 | |||
AK088614.1 | - | 2 | 175 | |||
AK160884.1 | - | 2 | 206 | |||
AK162755.1 | - | 2 | 212 | |||
AK172894.1 | - | 2 | 193 | |||
BC010489.1 | - | 2 | 198 | 4168214 |