Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019945.2 | NP_064329.2 | 7 | 831 | ||
GenBank mRNA | AF077818.1 | - | 7 | 814 | ||
AK141034.1 | - | 7 | 837 | |||
BC054524.1 | - | 7 | 831 | 6400705 |