Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019953.1 | NP_064337.1 | 2 | 415 | ||
GenBank mRNA | AF186115.1 | - | 2 | 415 | ||
AK007914.1 | - | 2 | 176 | |||
AK013014.1 | - | 2 | 286 | |||
AK013568.1 | - | 2 | 157 | |||
AK019927.1 | - | 2 | 287 | |||
AK150953.1 | - | 2 | 359 | |||
AK179967.1 | - | 2 | 115 | |||
AK180663.1 | - | 2 | 147 | |||
AK213625.1 | - | 2 | 212 | |||
BC008261.1 | - | 2 | 187 | 2650612 |