Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hacl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019975.3 | NP_064359.2 | 1, 2 | 170 | ||
GenBank mRNA | AJ132139.1 | - | 1, 2 | 168 | ||
AK005505.1 | - | 1, 2 | 157 | |||
AK041686.1 | - | 1, 2 | 166 | |||
AK050078.1 | - | 1, 2 | 170 | |||
AK161488.1 | - | 150 | ||||
BC021360.1 | - | 1, 2 | 101 | 5011451 | ||
BY340097.1 | - | 1, 2 | 170 |