Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019979.2 | NP_064363.2 | 4 | 348 | ||
GenBank mRNA | AF170920.1 | - | 4 | 377 | ||
AK003206.1 | - | 4 | 276 | |||
AK006674.1 | - | 4 | 248 | |||
AK007523.1 | - | 4 | 260 | |||
AK008419.1 | - | 4 | 284 | |||
BC096044.1 | - | 4 | 230 | 1248280 | ||
BC096677.1 | - | 4 | 278 | 874920 | ||
BY095166.1 | - | 4 | 348 |
Gene Symbol: BC052688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066226.1 | - | 2641 |
Gene Symbol: Gm12060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027278.1 | - | 1 | 322 |