Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019981.2 | NP_064365.1 | 3 | 235 | ||
GenBank mRNA | AB022914.1 | - | 3 | 233 | ||
AK005769.1 | - | 3 | 229 | |||
BC048475.1 | - | 3 | 226 | 6740387 | ||
DQ222038.1 | - | 434 | ||||
DQ222039.1 | - | 372 | ||||
DQ222040.1 | - | 735 | ||||
DQ222041.1 | - | 676 | ||||
DQ222042.1 | - | 3 | 237 |