Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163780.1 | NP_001157252.1 | 3, 4 | 624 | ||
NM_019987.2 | NP_064371.2 | 3, 4 | 552 | |||
GenBank mRNA | AF225918.1 | - | 3, 4 | 392 | ||
AK087484.1 | - | 3, 4 | 475 | |||
AK133537.1 | - | 3, 4 | 491 | |||
AK154185.1 | - | 3, 4 | 625 | |||
AK206365.1 | - | 2, 3 | 205 | |||
BC028863.1 | - | 2, 3 | 309 | 4224269 |