Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019990.4 | NP_064374.1 | 6, 7 | 785 | ||
GenBank mRNA | AB031550.1 | - | 7, 8 | 962 | ||
BC046335.1 | - | 6, 7 | 874 | |||
BC058773.1 | - | 6, 7 | 917 | |||
BC080808.1 | - | 6, 7 | 980 | |||
BC094007.1 | - | 6, 7 | 798 | |||
BC116984.1 | - | 5, 6 | 693 | 40126303 | ||
BC120641.1 | - | 5, 6 | 693 | 40129564 | ||
BC144877.1 | - | 4, 5 | 640 | |||
BG078855.2 | - | 5, 6 | 571 |