Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190320.1 | NP_001177249.1 | 2, 3 | 296 | ||
NM_020001.2 | NP_064385.1 | 3, 4 | 386 | |||
GenBank mRNA | AF240357.1 | - | 3, 4 | 355 | ||
AF240359.1 | - | 3, 4 | 355 | |||
AK089166.1 | - | 3, 4 | 373 | |||
AK131697.1 | - | 3, 4 | 373 | |||
AK154673.1 | - | 2, 3 | 283 | |||
AK171046.1 | - | 4, 5 | 556 | |||
BC023008.1 | - | 3, 4 | 355 | 5376247 |