Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019913.5 | NP_064297.1 | 4 | 552 | ||
GenBank mRNA | AK002358.1 | - | 4 | 527 | ||
AK010917.1 | - | 4 | 553 | |||
AK147164.1 | - | 3 | 1480 | |||
AK149855.1 | - | 4 | 536 | |||
AK167754.1 | - | 4 | 518 | |||
AK167925.1 | - | 4 | 490 | |||
AK168322.1 | - | 4 | 518 | |||
AK183615.1 | - | 1 | 36 | |||
BC068182.1 | - | 4 | 538 | 30633447 | ||
U85089.1 | - | 4 | 515 |