Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007332.2 | NP_015628.2 | 22 | 2798 | ||
XM_011517624.2 | XP_011515926.1 | 23 | 7614 | |||
XM_011517625.2 | XP_011515927.1 | 24 | 7773 | |||
XM_017013946.1 | XP_016869435.1 | 23 | 4076 | |||
GenBank mRNA | AK307942.1 | - | 22 | 2813 | ||
Y10601.1 | - | 22 | 2797 |