Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019925.4 | NP_064309.1 | 3 | 579 | ||
GenBank mRNA | AF083442.1 | - | 3 | 615 | ||
AK089866.1 | - | 3 | 556 | |||
AK153765.1 | - | 3 | 580 | |||
AK154509.1 | - | 3 | 601 | |||
AK157600.1 | - | 3 | 607 | |||
BC120522.1 | - | 2 | 506 | 40129445 | ||
BC120524.1 | - | 2 | 506 | 40129447 |
Set Membership: |
G-protein coupled receptor |