Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019953.1 | NP_064337.1 | 3 | 445 | ||
GenBank mRNA | AF186115.1 | - | 3 | 445 | ||
AK007914.1 | - | 3 | 206 | |||
AK013014.1 | - | 3 | 316 | |||
AK013568.1 | - | 3 | 187 | |||
AK019927.1 | - | 3 | 317 | |||
AK150953.1 | - | 3 | 389 | |||
AK179967.1 | - | 3 | 145 | |||
AK213625.1 | - | 3 | 242 | |||
BC008261.1 | - | 3 | 217 | 2650612 |