Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019956.1 | NP_064340.1 | 1 | 264 | ||
GenBank mRNA | AB033744.1 | - | 1 | 264 | ||
AB100413.1 | - | 1 | 264 | |||
AK003610.1 | - | 1 | 276 | |||
AK003704.1 | - | 1 | 276 | |||
AY033497.1 | - | 1 | 264 | |||
BC125346.1 | - | 1 | 259 | 40129861 | ||
BC125348.1 | - | 1 | 259 | 40129863 |