Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019957.4 | NP_064341.3 | 4, 5 | 711 | ||
GenBank mRNA | AF128888.1 | - | 3, 4 | 590 | ||
AK079224.1 | - | 4, 5 | 712 | |||
BC075661.1 | - | 3, 4 | 466 | 30606058 |