Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019979.2 | NP_064363.2 | 4, 5 | 423 | ||
GenBank mRNA | AF170920.1 | - | 4, 5 | 452 | ||
AK003206.1 | - | 4, 5 | 351 | |||
AK003280.1 | - | 17 | ||||
AK006674.1 | - | 4, 5 | 323 | |||
AK007523.1 | - | 4, 5 | 335 | |||
AK008419.1 | - | 4, 5 | 359 | |||
BC038049.1 | - | 4, 5 | 364 | 2645414 | ||
BC096044.1 | - | 4, 5 | 305 | 1248280 | ||
BC096677.1 | - | 4, 5 | 353 | 874920 |
Gene Symbol: BC052688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066226.1 | - | 2716 |
Gene Symbol: Gm12060 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027278.1 | - | 1 | 397 |