Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038010.2 | NP_001033099.1 | 3 | 626 | ||
NM_020004.5 | NP_064388.2 | 3 | 626 | |||
GenBank mRNA | AF254441.1 | - | 3 | 563 | ||
AK137358.1 | - | 3 | 603 | |||
AK145205.1 | - | 3 | 614 | |||
AK147042.1 | - | 3 | 576 | |||
AK150225.1 | - | 3 | 604 | |||
AK154866.1 | - | 3 | 614 | |||
AK155347.1 | - | 3 | 607 | |||
AK155791.1 | - | 3 | 627 | |||
AK155842.1 | - | 3 | 627 | |||
AK158079.1 | - | 3 | 614 | |||
AK185926.1 | - | 1 | 121 | |||
BC011088.1 | - | 3 | 332 | |||
BC051952.1 | - | 3 | 608 | |||
BC060050.1 | - | 3 | 608 | |||
BC063752.1 | - | 3 | 611 | 6333090 |