Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019875.2 | NP_063928.2 | 12 | 2693 | ||
GenBank mRNA | AB093298.1 | - | 12 | 2675 | ||
AF216495.1 | - | 12 | 2686 | |||
AK044140.1 | - | 12 | 2671 | |||
BC053014.1 | - | 12 | 2646 | 5686698 |