Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019921.3 | NP_064305.2 | 9, 10 | 1656 | ||
GenBank mRNA | AF021833.1 | - | 6, 7 | 1010 | ||
AK049399.1 | - | 9, 10 | 1625 | |||
AK051044.1 | - | 7, 8 | 1291 | |||
AK083255.1 | - | 9, 10 | 1625 | |||
AK162966.1 | - | 7, 8 | 1448 | |||
BC038483.1 | - | 6, 7 | 714 | |||
BC054105.1 | - | 6, 7 | 959 | 30022728 |