Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019924.2 | NP_064308.2 | 5 | 605 | ||
GenBank mRNA | AF074714.1 | - | 5 | 577 | ||
AK089467.1 | - | 5 | 596 | |||
AK154674.1 | - | 5 | 604 | |||
BC012964.1 | - | 5 | 615 | 4160873 |