Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301671.1 | NP_001288600.1 | 4 | 1425 | ||
NM_001301672.1 | NP_001288601.1 | 4 | 1257 | |||
NM_001301673.1 | NP_001288602.1 | 4 | 1308 | |||
NM_019929.4 | NP_064313.1 | 4 | 1299 | |||
GenBank mRNA | AF063847.1 | - | 4 | 1126 | ||
AK013019.1 | - | 4 | 1168 | |||
AK148306.1 | - | 4 | 1163 | |||
AK150063.1 | - | 4 | 1172 | |||
AK150200.1 | - | 4 | 1172 | |||
AK160169.1 | - | 4 | 1171 | |||
AK162035.1 | - | 4 | 1172 | |||
AK167207.1 | - | 4 | 1140 | |||
AK168988.1 | - | 4 | 1163 | |||
AK170896.1 | - | 4 | 1357 | |||
AK171169.1 | - | 4 | 1357 | |||
AK207570.1 | - | 1 | 195 |