Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161822.1 | NP_001155294.1 | 5 | 690 | ||
NM_019958.4 | NP_064342.1 | 4 | 620 | |||
GenBank mRNA | AF191555.1 | - | 4 | 562 | ||
AK018279.1 | - | 4 | 604 | |||
AK030492.1 | - | 4 | 621 | |||
AK045770.1 | - | 5 | 657 | |||
AK046009.1 | - | 4 | 579 | |||
AK078745.1 | - | 4 | 579 | |||
AK163188.1 | - | 4 | 577 |