Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019977.2 | NP_064361.2 | 10 | 855 | ||
GenBank mRNA | AF197127.1 | - | 10 | 853 | ||
AK002406.1 | - | 10 | 867 | |||
AK165481.1 | - | 1 | 2355 | |||
AY738257.1 | - | 9 | 825 | |||
BC013543.1 | - | 10 | 855 | 4242159 |