Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204239.1 | NP_001191168.1 | 4 | 685 | ||
NM_001204253.1 | NP_001191182.1 | 3 | 533 | |||
NM_019985.3 | NP_064369.1 | 5 | 781 | |||
GenBank mRNA | AF201457.1 | - | 5 | 492 | ||
AK007925.1 | - | 5 | 761 | |||
AK154961.1 | - | 36 | ||||
BC064054.1 | - | 5 | 781 | 574821 | ||
EF070191.1 | - | 4 | 375 | |||
EF070192.1 | - | 3 | 220 |