Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019992.3 | NP_064376.1 | 8 | 808 | ||
GenBank mRNA | AB036058.1 | - | 8 | 798 | ||
AK041474.1 | - | 8 | 809 | |||
AK153661.1 | - | 8 | 887 | |||
AK162655.1 | - | 8 | 890 | |||
BC145550.1 | - | 7 | 710 | 9054114 | ||
BC145828.1 | - | 8 | 823 | 40131131 | ||
BC145830.1 | - | 8 | 824 | 40131133 |