Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003719.2 | NP_001003719.1 | 33 | 5252 | ||
NM_001112714.2 | NP_001106185.1 | 33 | 5252 | |||
NM_001286263.1 | NP_001273192.1 | 34 | 5393 | |||
NM_019994.5 | NP_064378.4 | 33 | 5252 | |||
GenBank mRNA | AB032400.1 | - | 6 | 692 | ||
AK083518.1 | - | 5 | 846 | |||
AK083746.1 | - | 5 | 846 | |||
AY066011.1 | - | 21 | 2838 | |||
AY596972.1 | - | 33 | 4903 | |||
AY596973.1 | - | 33 | 4903 | |||
AY596974.1 | - | 33 | 4903 | |||
BC145119.1 | - | 34 | 4974 | 9053667 | ||
BC150707.1 | - | 34 | 4974 | 9087618 |