Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282470.2 | NP_001269399.1 | 2, 3 | 425 | ||
NM_001282473.2 | NP_001269402.1 | 3, 4 | 533 | |||
NM_014619.4 | NP_055434.2 | 3, 4 | 385 | |||
XM_011542784.1 | XP_011541086.1 | 2, 3 | 243 | |||
XM_017017621.1 | XP_016873110.1 | 2, 3 | 243 | |||
XM_017017622.1 | XP_016873111.1 | 2, 3 | 243 | |||
GenBank mRNA | AK292726.1 | - | 3, 4 | 372 | ||
AK314284.1 | - | 2, 3 | 427 | |||
BC146652.1 | - | 3, 4 | 483 | 40147575 | ||
BC150173.1 | - | 3, 4 | 483 | 40147389 | ||
DA246271.1 | - | 3, 4 | 383 | |||
DA740816.1 | - | 3, 4 | 375 | |||
S67803.1 | - | 1, 2 | 85 |