Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038010.2 | NP_001033099.1 | 16 | 2231 | ||
NM_020004.5 | NP_064388.2 | 16 | 2234 | |||
GenBank mRNA | AF254441.1 | - | 16 | 2171 | ||
AK145205.1 | - | 14 | 2231 | |||
AK147042.1 | - | 16 | 2179 | |||
AK150225.1 | - | 16 | 2212 | |||
AK154866.1 | - | 15 | 2309 | |||
AK155347.1 | - | 16 | 2408 | |||
AK155791.1 | - | 16 | 2233 | |||
AK155842.1 | - | 16 | 2232 | |||
AK158079.1 | - | 16 | 2222 | |||
AK179662.1 | - | 4 | 284 | |||
BC003983.1 | - | 1 | 29 | |||
BC011088.1 | - | 16 | 2141 | |||
BC051952.1 | - | 14 | 2455 | |||
BC060050.1 | - | 14 | 2455 | |||
BC063752.1 | - | 16 | 2216 | 6333090 |