Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020009.2 | NP_064393.2 | 6 | 858 | ||
GenBank mRNA | AF152838.1 | - | 6 | 833 | ||
AK143381.1 | - | 3 | 269 | |||
AK172584.1 | - | 6 | 849 | |||
BC112904.1 | - | 6 | 816 | 6465126 |