Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015166.3 | NP_055981.1 | 10 | 1419 | ||
NM_139202.2 | NP_631941.1 | 10 | 1058 | |||
XM_011530678.1 | XP_011528980.1 | 10 | 1180 | |||
XM_017028671.1 | XP_016884160.1 | 9 | 847 | |||
GenBank mRNA | AF319633.1 | - | 10 | 981 | ||
AK124264.1 | - | 10 | 1420 | |||
AK294048.1 | - | 8 | 809 | |||
AK295106.1 | - | 9 | 680 | |||
AK297341.1 | - | 9 | 779 | |||
AK299841.1 | - | 9 | 827 | |||
BC028425.2 | - | 10 | 1018 | 4817175 | ||
BC070042.1 | - | 10 | 1016 | 5285102 | ||
CR456460.1 | - | 9 | 837 | |||
D25217.3 | - | 10 | 1095 |