Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020015.2 | NP_064399.1 | 1 | 962 | ||
GenBank mRNA | BC116904.1 | - | 1 | 962 | 40126223 | |
CJ223782.1 | - | 1 | 77 |