Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magea6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020019.3 | NP_064403.1 | 2 | 555 | ||
GenBank mRNA | BC109339.2 | - | 2 | 566 | 40052528 | |
BC116902.1 | - | 1 | 465 | 40126221 |