Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278926.1 | NP_001265855.1 | 1 | 200 | ||
NM_001278927.1 | NP_001265856.1 | 1 | 200 | |||
NM_001278929.1 | NP_001265858.1 | 1 | 200 | |||
NM_015342.3 | NP_056157.1 | 1 | 203 | |||
XM_005248474.3 | XP_005248531.1 | 1 | 143 | |||
XM_006714583.3 | XP_006714646.1 | 1 | 96 | |||
GenBank mRNA | AK025679.1 | - | 1 | 150 | ||
AK093675.1 | - | 1 | 145 | |||
AK298175.1 | - | 1 | 140 | |||
AK300022.1 | - | 1 | 143 | |||
AK301654.1 | - | 1 | 140 | |||
BC015385.1 | - | 1 | 131 | 4390706 | ||
BX642958.1 | - | 1 | 208 | |||
D38552.1 | - | 1 | 126 | |||
DB454273.1 | - | 1 | 200 |