Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scube2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020052.2 | NP_064436.2 | 4 | 564 | ||
GenBank mRNA | AK142950.1 | - | 4 | 530 |