Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145960.1 | NP_001139432.1 | 4 | 641 | ||
NM_020258.4 | NP_064654.3 | 4 | 641 | |||
GenBank mRNA | AF121081.1 | - | 4 | 540 | ||
AK028984.1 | - | 4 | 317 | |||
AK031045.1 | - | 4 | 579 | |||
AK079414.1 | - | 4 | 317 | |||
AK079823.1 | - | 4 | 642 | |||
AK220151.1 | - | 2 | 84 | |||
BC038000.1 | - | 4 | 312 | |||
BC063326.1 | - | 4 | 329 | 30301217 |