Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146200.1 | NP_001139672.1 | 2 | 462 | ||
NM_001146201.1 | NP_001139673.1 | 2 | 518 | |||
NM_020272.2 | NP_064668.2 | 2 | 639 | |||
GenBank mRNA | AF208345.1 | - | 1 | 318 | ||
AJ249280.1 | - | 2 | 639 | |||
AK040543.1 | - | 2 | 424 | |||
AK132553.1 | - | 2 | 464 | |||
AK137470.1 | - | 2 | 371 | |||
AK137620.1 | - | 2 | 520 | |||
BC051246.1 | - | 2 | 379 | 6514979 |