Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020284.1 | NP_064680.1 | 2, 3 | 206 | ||
GenBank mRNA | AF245399.1 | - | 2, 3 | 206 | ||
AK005429.1 | - | 2, 3 | 226 | |||
AK014432.1 | - | 2, 3 | 222 | |||
AY014778.1 | - | 2, 3 | 165 | |||
AY943668.1 | - | 2, 3 | 275 | |||
BC100338.1 | - | 2, 3 | 201 | 30712889 |